Learn More
Medchemexpress LLC HY-15826 10mg , SGC-CBP30 CAS:1613695-14-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1582610MG
Encompass_Preferred
HY-15826 10mg SGC-CBP30 CAS: 1613695-14-9 SGC-CBP30 is a potent and highly selective CBP/p300 bromodomain (Kds of 21 nM and 32 nM for CBP and p300, respectively) inhibitor, displaying 40-fold selectivity over the first bromodomain of BRD4 [BRD4(1)] bound. SGC-CBP30 strongly reduces secretion of IL-17A in Th17 cells and has anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9760