Learn More
Apexbio Technology LLC SGC-CBP30 1613695-14-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061504
Encompass_Preferred
SGC-CBP30 (CAS 1613695-14-9) is a selective small-molecule inhibitor targeting the bromodomains of CREB-binding protein (CREBBP) and E1A-binding protein p300 (EP300) exhibiting IC50 values of 21 nM and 38 nM respectively Both EP300 and CREBBP function as transcriptional coactivators involved in diverse regulatory processes including cell proliferation differentiation and transcription factor activation Through its inhibitory action SGC-CBP30 is utilized widely in biomedical research to investigate the molecular roles of CREBBP/EP300 as well as in studies examining related signaling pathways in tumor biology and cellular regulation
Catalog No. 50-000-61504 Supplier Apexbio Technology LLC Supplier No. A44915.1
SGC-CBP30 (CAS 1613695-14-9) is a selective small-molecule inhibitor targeting the bromodomains of CREB-binding protein (CREBBP) and E1A-binding protein p300 (EP300) exhibiting IC50 values of 21 nM and 38 nM respectively Both EP300 and CREBBP function as transcriptional coactivators involved in diverse regulatory processes including cell proliferation differentiation and transcription factor activation Through its inhibitory action SGC-CBP30 is utilized widely in biomedical research to investigate the molecular roles of CREBBP/EP300 as well as in studies examining related signaling pathways in tumor biology and cellular regulation