Learn More
Medchemexpress LLC HY-112804 10mg Medchemexpress, SGC-iMLLT CAS:2255338-25-9 Purity:>98%
SDP
Catalog No. 501969176
Encompass_Preferred
Medchemexpress, HY-112804 10mg SGC-iMLLT CAS:2255338-25-9 SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9176 Supplier Medchemexpress LLC Supplier No. HY11280410MG
Medchemexpress, HY-112804 10mg SGC-iMLLT CAS:2255338-25-9 SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.