Learn More
Medchemexpress LLC HY-112804 5mg Medchemexpress, SGC-iMLLT CAS:2255338-25-9 Purity:>98%
SDP
Catalog No. 501932438
Encompass_Preferred
Medchemexpress, HY-112804 5mg SGC-iMLLT CAS:2255338-25-9 SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC 50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with K d s of 0.129 and 0.077 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2438 Supplier Medchemexpress LLC Supplier No. HY1128045MG
Medchemexpress, HY-112804 5mg SGC-iMLLT CAS:2255338-25-9 SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC 50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with K d s of 0.129 and 0.077 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.