Learn More
Cayman Chemical SGC Epigenetic Probe Set, 1 ea
SDP
Catalog No. 502966801
Encompass_Preferred
Contains a collection of small, drug-like molecules that inhibit or antagonize epigenetic readers, writers, and erasers each packaged as a 1 mg sample size for ease of preclinical target validation during early stages of drug discovery
Catalog No. 50-296-6801 Supplier Cayman Chemical Supplier No. 177481 EA
Contains a collection of small, drug-like molecules that inhibit or antagonize epigenetic readers, writers, and erasers each packaged as a 1 mg sample size for ease of preclinical target validation during early stages of drug discovery