Learn More
Cayman Chemical SGC2085hydrochlorIde 10mg
SDP
Catalog No. 502986674
Encompass_Preferred
A PRMT4/CARM1 inhibitor (IC50 = 50 nM); highly selective for PRMT4/CARM1 over other PRMTs; weakly inhibits PRMT6 (IC50 = 5.2 mM)
Catalog No. 50-298-6674 Supplier Cayman Chemical Supplier No. 2187910 MG
A PRMT4/CARM1 inhibitor (IC50 = 50 nM); highly selective for PRMT4/CARM1 over other PRMTs; weakly inhibits PRMT6 (IC50 = 5.2 mM)