Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SGC3027 | 2624313-13-7 | 98.1% | 787.37 | 10 MG
SDP

Catalog No. 5000437468
Encompass_Preferred

SGC3027 is a histone methyltransferase inhibitor and is identified as the first potent, selective, and cell-active chemical probe for PRMT7. It acts as a pro-drug of SGC8158, releasing its active component upon reduction by reductases within the cell. In cellular assays using C2C12 cells, SGC3027 inhibits the methylation of HSP70 with an IC50 of 1.3 μM.

  • Target: PRMT7.
  • Mechanism: Histone methyltransferase inhibitor.
  • Cellular activity: Inhibits the methylation of HSP70.
  • Appearance: Solid.
  • Color: Light yellow to yellow.
  • Solubility: Soluble in DMSO (250 mg/mL).

Catalog No. 50-004-37468 Supplier Medchemexpress LLC Supplier No. HY11244510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SGC3027 is a histone methyltransferase inhibitor and is identified as the first potent, selective, and cell-active chemical probe for PRMT7. It acts as a pro-drug of SGC8158, releasing its active component upon reduction by reductases within the cell. In cellular assays using C2C12 cells, SGC3027 inhibits the methylation of HSP70 with an IC50 of 1.3 μM.

  • Target: PRMT7.
  • Mechanism: Histone methyltransferase inhibitor.
  • Cellular activity: Inhibits the methylation of HSP70.
  • Appearance: Solid.
  • Color: Light yellow to yellow.
  • Solubility: Soluble in DMSO (250 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.