Learn More
Medchemexpress LLC SGC3027 | 2624313-13-7 | 98.1% | 787.37 | 50 MG
SDP
Catalog No. 5000438660
Encompass_Preferred
SGC3027 is a histone methyltransferase inhibitor, described as the first potent, selective, and cell-active chemical probe for PRMT7. It functions as a prodrug of SGC8158, becoming active upon cellular reduction by reductases. In C2C12 cellular assays, SGC3027 inhibits the methylation of HSP70 with an IC50 of 1.3 μM.
  • Potent, selective, and cell-active chemical probe for PRMT7
  • Prodrug that releases its active component (SGC8158) upon cellular reduction
Catalog No. 50-004-38660 Supplier Medchemexpress LLC Supplier No. HY11244550MG
SGC3027 is a histone methyltransferase inhibitor, described as the first potent, selective, and cell-active chemical probe for PRMT7. It functions as a prodrug of SGC8158, becoming active upon cellular reduction by reductases. In C2C12 cellular assays, SGC3027 inhibits the methylation of HSP70 with an IC50 of 1.3 μM.
  • Potent, selective, and cell-active chemical probe for PRMT7
  • Prodrug that releases its active component (SGC8158) upon cellular reduction