Learn More
Medchemexpress LLC HY-112445 5mg , SGC3027 CAS:2624313-13-7 Purity:>98%
SDP
Catalog No. 502261664
Encompass_Preferred
HY-112445 5mg SGC3027 CAS: 2624313-13-7 SGC3027 is a histone methyltransferase inhibitor. SGC3027 is the first potent, selective and cell active chemical probe for PRMT7. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1664 Supplier Medchemexpress LLC Supplier No. HY1124455MG
HY-112445 5mg SGC3027 CAS: 2624313-13-7 SGC3027 is a histone methyltransferase inhibitor. SGC3027 is the first potent, selective and cell active chemical probe for PRMT7. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.