Learn More
Biotium SGC5
Supplier:  Biotium 70057
Encompass_Preferred
SGC5 is a coumarin styryl dye that binds to membranes in a manner similar to the FM dyes. It becomes highly fluorescent upon insertion into lipid bilayers. Properties: Ex/Em 484/563 nm (in MeOH); Dark Orange solid soluble in water.FM is a registered trademark of Thermo Fisher Scientific.
Catalog No. 50-196-4764