Learn More
Boca Scientific Inc Dri-Capsules Buccal DNA Stabilization Capsules for use with SK-1S Buccal Swabs, Pack of 50 with 50 Swabs included
Catalog No. NC0911135
Encompass
The SGC-50/SK1 kit includes 50 SK-1S buccal swabs and 50 Dri-Capsules for efficient DNA collection and stabilization. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. The shaft has a break point near the swab head to easily snap off the swab head into the 5ml tube. The Dri-Capsules conveniently stabilize DNA on SK-1S swabs for over 3 years at room temperature. After obtaining the sample, the capsule is inserted into the 5ml collection tube along with the used swab head and the tube is sealed with the provided cap. The capsules have a non-toxic coating containing a high-grade silica gel with a color indicator to display continued active stabilization.
Catalog No. NC0911135 Supplier Boca Scientific Inc Supplier No. SGC50SK1
The SGC-50/SK1 kit includes 50 SK-1S buccal swabs and 50 Dri-Capsules for efficient DNA collection and stabilization. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. The shaft has a break point near the swab head to easily snap off the swab head into the 5ml tube. The Dri-Capsules conveniently stabilize DNA on SK-1S swabs for over 3 years at room temperature. After obtaining the sample, the capsule is inserted into the 5ml collection tube along with the used swab head and the tube is sealed with the provided cap. The capsules have a non-toxic coating containing a high-grade silica gel with a color indicator to display continued active stabilization.