Learn More
Active Motif SGC707
SDP
Supplier:  Active Motif 14131
Encompass
Active Motif's SGC707 is a potent and selective allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3), IC50 = 31 nM and selective against 31 other methyltransferases. SGC707 is an extremely useful probe for investigating the role of PRMT3 in normal and pathophysiology. it is cell permeable and active in vivo.
Catalog No. NC1841458