Learn More
Teknova SGCAA Media with Galactose. 1000mL, Sterile.
SDP
Catalog No. 501907854
Encompass
Synthetic Defined Media (SDM) is a type of growth media that is formulated to provide cells or tissues with the nutrients they need to thrive. This media is used in research labs to study the effects of various chemicals on cells. SDM allows for more control over the environment in which cells are grown, and as a result, can provide insights that would be otherwise difficult to obtain.
This item is not returnable. View return policy
Catalog No. 50-190-7854 Supplier Teknova Supplier No. 2S0542
Synthetic Defined Media (SDM) is a type of growth media that is formulated to provide cells or tissues with the nutrients they need to thrive. This media is used in research labs to study the effects of various chemicals on cells. SDM allows for more control over the environment in which cells are grown, and as a result, can provide insights that would be otherwise difficult to obtain.