Learn More
Medchemexpress LLC HY-101127 1mg , SGD-1882 CAS:1222490-34-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011271MG
Encompass_Preferred
HY-101127 1mg SGD-1882 CAS: 1222490-34-7 SGD-1882 is a cytotoxic, DNA minor-groove crosslinking agent pyrrolobenzodiazepine (PBD) dimer, acting as the payload for ADCs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9876