Learn More
Trajan Scientific SGE FORTE BPX-VOLATILES
SDP
Catalog No. 502530226
Encompass_Preferred
SGE GC Capillary Columns: Cyanopropyl Polysilphenylene, Diameter Inner: 0.25 mm, Film Thickness: 1.4 um, Particle Size: 1.4 um, For Use With: Alcohol Analysis, Simulated Distillation, Volatile Organic Compounds Analysis, Length: 60 m, Packing Material: BPX-Volatiles
Catalog No. 50-253-0226 Supplier Trajan Scientific Supplier No. 054981
SGE GC Capillary Columns: Cyanopropyl Polysilphenylene, Diameter Inner: 0.25 mm, Film Thickness: 1.4 um, Particle Size: 1.4 um, For Use With: Alcohol Analysis, Simulated Distillation, Volatile Organic Compounds Analysis, Length: 60 m, Packing Material: BPX-Volatiles