Learn More
Trajan Scientific SGE FORTE GC COLUMN BPX-1
SDP
Catalog No. 502530230
Encompass_Preferred
SGE GC Capillary Columns: Dimethyl Polysiloxane, Diameter Inner: 0.53 mm, Film Thickness: 0.9 um, Length: 10 m, Temperature Range: -30 deg.C to +400 deg.C, For Use With: Simulated Distillation, Packing Material: BPX1, Particle Size: 0.9 um, Stationary Phase: 100% Dimethyl Polysiloxane
Catalog No. 50-253-0230 Supplier Trajan Scientific Supplier No. 054801
SGE GC Capillary Columns: Dimethyl Polysiloxane, Diameter Inner: 0.53 mm, Film Thickness: 0.9 um, Length: 10 m, Temperature Range: -30 deg.C to +400 deg.C, For Use With: Simulated Distillation, Packing Material: BPX1, Particle Size: 0.9 um, Stationary Phase: 100% Dimethyl Polysiloxane