Learn More
Trajan Scientific SGE FORTE GC COLUMN HT-5
SDP
Catalog No. 502530235
Encompass_Preferred
SGE GC Capillary Columns: Phenyl (equiv.) Polycarborane Siloxane, Diameter Inner: 0.53 mm, Film Thickness: 0.15 um, Length: 12 m, Polyimide Clad, For Use With: Simulated Distillation, Pesticide Analysis, Herbicides Analysis, Packing Material: HT5, Particle Size: 0.15 um
Catalog No. 50-253-0235 Supplier Trajan Scientific Supplier No. 054657
SGE GC Capillary Columns: Phenyl (equiv.) Polycarborane Siloxane, Diameter Inner: 0.53 mm, Film Thickness: 0.15 um, Length: 12 m, Polyimide Clad, For Use With: Simulated Distillation, Pesticide Analysis, Herbicides Analysis, Packing Material: HT5, Particle Size: 0.15 um