Learn More
New Era Pump Systems Inc SGE PTFE Tipped Plunger Gas Tight Glass Syringe, 500ul, Fixed Metal Luer Lock, 400psi
Catalog No. NC3824950
Encompass
A PTFE tip plunger minimizes carryover and prevents particulate matter from getting between the plunger and barrel by effectively wiping the barrel inner diameter during the plunger stroke. PTFE tipped plungers are suitable for both liquid and gas samples. Individually boxed. Pistons and barrels are interchangeable.
Catalog No. NC3824950 Supplier New Era Pump Systems Inc Supplier No. SYRGL500MCLLS
A PTFE tip plunger minimizes carryover and prevents particulate matter from getting between the plunger and barrel by effectively wiping the barrel inner diameter during the plunger stroke. PTFE tipped plungers are suitable for both liquid and gas samples. Individually boxed. Pistons and barrels are interchangeable.