Learn More
World Precision Instrument SGE Syringes with Replaceable Beveled Needles-SGE001RN
SDP
Catalog No. 502538586
Encompass
A 20° bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.
Catalog No. 50-253-8586 Supplier World Precision Instrument Supplier No. SGE001RN
A 20° bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.