Learn More
World Precision Instrument SGE Syringes with Replaceable Beveled Needles-SGE100RN
SDP
Catalog No. 502538581
Encompass
A 20° bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.
Catalog No. 50-253-8581 Supplier World Precision Instrument Supplier No. SGE100RN
A 20° bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.