Learn More
Cayman Chemical SGernylgernylLglutathIon 10mg
SDP
Catalog No. 502892845
Encompass_Preferred
A P2RY8 ligand; selectively induces internalization of P2RY8 over S1P2, GPR55, CysLT1 receptor, and CysLT2 receptor at 100 nM; inhibits CXCL12-induced migration of P2RY8-expressing WEHI-231 B cell lymphoma cells and isolated human tonsil germinal center B cells at 10 nM
Catalog No. 50-289-2845 Supplier Cayman Chemical Supplier No. 900411610 MG
A P2RY8 ligand; selectively induces internalization of P2RY8 over S1P2, GPR55, CysLT1 receptor, and CysLT2 receptor at 100 nM; inhibits CXCL12-induced migration of P2RY8-expressing WEHI-231 B cell lymphoma cells and isolated human tonsil germinal center B cells at 10 nM