Learn More
Selleck Chemical LLC SGI-1027 10mg 1020149-73-8 DNA Methyltransferase Inhibitor II
SDP
Supplier:  Selleck Chemical LLC S727610MG
Encompass_Preferred
SGI-1027 (DNA Methyltransferase Inhibitor II) is a DNMT inhibitor with IC50 of 6, 8, 7.5 ?M for DNMT1, DNMT3A, and DNMT3B in cell-free assays, respectively. SGI?1027 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4586