Learn More
Selleck Chemical LLC SGI-1027
SDP
Catalog No. 5000004604
Encompass_Preferred
SGI-1027 (DNA Methyltransferase Inhibitor II) is a DNMT inhibitor with IC50 of 6 8 7 5 M for DNMT1 DNMT3A and DNMT3B in cell-free assays respectively SGI 1027 induces apoptosis
Catalog No. 50-000-04604 Supplier Selleck Chemical LLC Supplier No. S7276100MG
SGI-1027 (DNA Methyltransferase Inhibitor II) is a DNMT inhibitor with IC50 of 6 8 7 5 M for DNMT1 DNMT3A and DNMT3B in cell-free assays respectively SGI 1027 induces apoptosis