Learn More
TARGETMOL CHEMICALS INC SGI-1027 25MG
SDP
Catalog No. 502581058
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SGI-1027 (DNA Methyltransferase Inhibitor II) is an effective and selective inhibitor of DNA methyltransferase (DNMT). The IC50 of SGI-1027(SGI1027) against DNMT1 DNMT3A and DNMT3B are 6 8 7.5 uM respectively. purity: 99%
Catalog No. 50-258-1058 Supplier TARGETMOL CHEMICALS INC Supplier No. T190425MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SGI-1027 (DNA Methyltransferase Inhibitor II) is an effective and selective inhibitor of DNA methyltransferase (DNMT). The IC50 of SGI-1027(SGI1027) against DNMT1 DNMT3A and DNMT3B are 6 8 7.5 uM respectively. purity: 99%