Learn More
Apexbio Technology LLC SGI-1776 free base 1025065-69-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061427
Encompass_Preferred
SGI-1776 free base (CAS 1025065-69-3) is an ATP-competitive inhibitor targeting Pim kinases a serine/threonine protein kinase family implicated in regulating cellular survival pathways It selectively inhibits all three isoforms Pim-1 Pim-2 and Pim-3 leading to decreased phosphorylation of downstream substrates involved in cell cycle regulation and apoptosis such as p21Cip1/WAF1 and Bad Cellular studies indicate that SGI-1776 induces G1 cell cycle arrest promotes apoptosis and overcomes chemoresistance mediated by multidrug resistance protein 1 (MDR1) in taxane-resistant prostate cancer cells
Catalog No. 50-000-61427 Supplier Apexbio Technology LLC Supplier No. A41925.1
SGI-1776 free base (CAS 1025065-69-3) is an ATP-competitive inhibitor targeting Pim kinases a serine/threonine protein kinase family implicated in regulating cellular survival pathways It selectively inhibits all three isoforms Pim-1 Pim-2 and Pim-3 leading to decreased phosphorylation of downstream substrates involved in cell cycle regulation and apoptosis such as p21Cip1/WAF1 and Bad Cellular studies indicate that SGI-1776 induces G1 cell cycle arrest promotes apoptosis and overcomes chemoresistance mediated by multidrug resistance protein 1 (MDR1) in taxane-resistant prostate cancer cells