Learn More
Selleck Chemical LLC SGI-1776 free base S2198-10mM/1mL
SDP
Catalog No. 503112847
Encompass_Preferred
SGI-1776 free base is a novel ATP competitive inhibitor of Pim1 with IC50 of 7 nM in a cell-free assay 50- and 10-fold selective versus Pim2 and Pim3 also potent to Flt3 and haspin SGI-1776 induces apoptosis and autophagy
Catalog No. 50-311-2847 Supplier Selleck Chemical LLC Supplier No. S219810MM/1ML
SGI-1776 free base is a novel ATP competitive inhibitor of Pim1 with IC50 of 7 nM in a cell-free assay 50- and 10-fold selective versus Pim2 and Pim3 also potent to Flt3 and haspin SGI-1776 induces apoptosis and autophagy