Learn More
Abcam SGI1776, 5MG
Catalog No. 502682930
Encompass_Preferred
MW 405.4 Da, Purity >99%. Novel ATP competitive inhibitor of Pim1 with IC50 of 7 nM, 50- and 10-fold selective versus Pim2 and Pim3. Preliminary results from studies treating prostate cancer cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2930 Supplier Abcam Supplier No. AB2735325MG
MW 405.4 Da, Purity >99%. Novel ATP competitive inhibitor of Pim1 with IC50 of 7 nM, 50- and 10-fold selective versus Pim2 and Pim3. Preliminary results from studies treating prostate cancer cells.
The product is subject to the following: Abcam Restricted Use Statement