Learn More
Sino Biological Recombinant Human RPS6KL1 Protein (Full Length, GST Tag)
Catalog No. 502573212
Encompass_Preferred
Recombinant full length human p70S6KL1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3212 Supplier Sino Biological Supplier No. R2534G1000
Recombinant full length human p70S6KL1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.