Learn More
Medchemexpress LLC HY-18607 5mg Medchemexpress, SGK1-IN-1 CAS:1279829-87-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY186075MG
Encompass_Preferred
Medchemexpress, HY-18607 5mg SGK1-IN-1 CAS:1279829-87-6 SGK1-IN-1 is a highly active and selective inhibitor of SGK-1, with an IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0009