Learn More
Medchemexpress LLC SGK1-IN-3 hydrochloride | 00-00-0 | 96.7% | 567.88 g/mol | C23H21Cl3N6O3S | 5 MG
SDP
Catalog No. 5000220984
Encompass_Preferred
SGK1-IN-3 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of serum- and glucocorticoid-regulated kinase 1 (SGK1). It has reported potent activity (IC50 <1 μM) and is supplied as a solid research reagent for biochemical and cellular studies, including exploratory research in osteoarthritis.
  • Potent SGK1 inhibition (IC50 <1 μM).
  • Supplied as a solid suitable for biochemical and cellular assays.
  • High reported purity for reliable experimental results.
  • Available in small research pack sizes for screening and optimization.
  • Useful for exploratory studies of SGK1-related pathways and disease models.
Catalog No. 50-002-20984 Supplier Medchemexpress LLC Supplier No. HY142686A5MG
SGK1-IN-3 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of serum- and glucocorticoid-regulated kinase 1 (SGK1). It has reported potent activity (IC50 <1 μM) and is supplied as a solid research reagent for biochemical and cellular studies, including exploratory research in osteoarthritis.
  • Potent SGK1 inhibition (IC50 <1 μM).
  • Supplied as a solid suitable for biochemical and cellular assays.
  • High reported purity for reliable experimental results.
  • Available in small research pack sizes for screening and optimization.
  • Useful for exploratory studies of SGK1-related pathways and disease models.