Learn More
Medchemexpress LLC SGK1-IN-3 hydrochloride | 00-00-0 | 96.7% | C23H21Cl3N6O3S | 10MG
SDP
Catalog No. 5000261118
Encompass_Preferred
SGK1-IN-3 hydrochloride is a research-grade small-molecule inhibitor of serum- and glucocorticoid-regulated kinase 1 (SGK1), reported to inhibit SGK1 with an IC50 of <1 μM. Supplied as the hydrochloride salt, it is intended for biochemical and cell-based research applications, including studies of signaling pathways related to osteoarthritis.
  • Potent SGK1 inhibition (IC50 <1 μM).
  • Hydrochloride salt form for improved solubility.
  • Suitable for biochemical and cell-based assays.
  • High reported purity (~96.7%).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-61118 Supplier Medchemexpress LLC Supplier No. HY142686A10MG
SGK1-IN-3 hydrochloride is a research-grade small-molecule inhibitor of serum- and glucocorticoid-regulated kinase 1 (SGK1), reported to inhibit SGK1 with an IC50 of <1 μM. Supplied as the hydrochloride salt, it is intended for biochemical and cell-based research applications, including studies of signaling pathways related to osteoarthritis.
  • Potent SGK1 inhibition (IC50 <1 μM).
  • Hydrochloride salt form for improved solubility.
  • Suitable for biochemical and cell-based assays.
  • High reported purity (~96.7%).
  • Available in small research pack sizes for laboratory use.