Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SGK1-IN-3 hydrochloride | 96.65% | C23H21Cl3N6O3S | 50 MG
SDP

Catalog No. 5000353714
Encompass_Preferred

SGK1-IN-3 hydrochloride (compound 3a) is a potent SGK1 inhibitor with an IC50 of <1 μM, and it has the potential for the research of osteoarthritis. This product is for research use only and not for sale to patients.

  • Potent SGK1 inhibitor
  • Potential for osteoarthritis research
  • Purity of 96.65%
  • Solid appearance with light yellow to yellow color
  • Molecular weight of 567.88
  • Soluble in DMSO at 125 mg/mL

Catalog No. 50-003-53714 Supplier Medchemexpress LLC Supplier No. HY142686A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SGK1-IN-3 hydrochloride (compound 3a) is a potent SGK1 inhibitor with an IC50 of <1 μM, and it has the potential for the research of osteoarthritis. This product is for research use only and not for sale to patients.

  • Potent SGK1 inhibitor
  • Potential for osteoarthritis research
  • Purity of 96.65%
  • Solid appearance with light yellow to yellow color
  • Molecular weight of 567.88
  • Soluble in DMSO at 125 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.