Learn More
Eurofins Discoverx SGK1 (S422D), unactive, 25 µg. (14-332)
Supplier:  Eurofins Discoverx 14332
Encompass
N-truncated (missing N-terminal 59 amino acids), N-terminal 6xHis-tagged recombinant, human SGK1 fusion protein, containing an S422D mutation. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8765