Learn More
Eurofins Discoverx SGK3, active, 10 µg. (14-647)
Supplier:  Eurofins Discoverx 14647
Encompass
N-terminal 6xHis-tagged recombinant, human SGK3 amino acids 119-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9115