Learn More
Aviva Systems Biology Corp SGK3 Antibody - N-terminal region (ARP55612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SGK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SGK3. Alternate names: CISK, DKFZp781N0293, SGK2, SGKLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61553