Learn More
Carnabio Usa Inc SGK3(SGKL)/5UG/TRUNCATED
Supplier:  Carnabio Usa Inc 011605
Encompass
SGK3(SGKL), Truncated protein, Active mutant [S486D], Amino Acid 119-496, NP_037389.4, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0622