Learn More
Euthanex Single Animal Anesthesia Machine with New TEC 3 Vaporizer - Key Fill
Catalog No. 501540149
Encompass_Preferred
Single Animal Anesthesia Machine with New TEC 3 Vaporizer - Key Fill. Includes two selectable outputs for an induction chamber and breather. Your choice of either Isoflurane OR Sevoflurane vaporizer with a "Key Fill" adaptor.
Catalog No. 50-154-0149 Supplier Euthanex Supplier No. EZ108SAKF
Single Animal Anesthesia Machine with New TEC 3 Vaporizer - Key Fill. Includes two selectable outputs for an induction chamber and breather. Your choice of either Isoflurane OR Sevoflurane vaporizer with a "Key Fill" adaptor.