Learn More
Euthanex Single Animal Anesthesia Machine - No Vaporizer
Catalog No. 501540150
Encompass_Preferred
Single Animal Anesthesia Machine - No Vaporizer. Includes two selectable outputs for an induction chamber and breather. Ready to go for many "Cage Mount" TEC 3 Vaporizers of your choice.
Catalog No. 50-154-0150 Supplier Euthanex Supplier No. EZ108SANV
Single Animal Anesthesia Machine - No Vaporizer. Includes two selectable outputs for an induction chamber and breather. Ready to go for many "Cage Mount" TEC 3 Vaporizers of your choice.