Learn More
Euthanex Single Animal Anesthesia System with Vaporizer, Mouse and Rat Breathers, Induction Chamber, Surgery Bed, Charcoal Filters and more.
Catalog No. 501540183
Encompass_Preferred
Single Animal Anesthesia System with TEC 3 Vaporizer and featuring two selectable outputs for an induction chamber and breather. Your choice of either Isoflurane or Sevoflurane vaporizer. Comes complete with Anesthesia Machine, Microflex Breather Base with (1) Nosecone for mouse & (1) Nosecone for rat, Sure-Seal Mouse/Rat Induction Chamber, (8) ReFresh Charcoal Canister FIlters, Oxygen Regulator for 'H' Tanks, Nosecone plugs, Plastic Surgery Bed and all connecting hoses.
Catalog No. 50-154-0183 Supplier Euthanex Supplier No. EZSA800
Single Animal Anesthesia System with TEC 3 Vaporizer and featuring two selectable outputs for an induction chamber and breather. Your choice of either Isoflurane or Sevoflurane vaporizer. Comes complete with Anesthesia Machine, Microflex Breather Base with (1) Nosecone for mouse & (1) Nosecone for rat, Sure-Seal Mouse/Rat Induction Chamber, (8) ReFresh Charcoal Canister FIlters, Oxygen Regulator for 'H' Tanks, Nosecone plugs, Plastic Surgery Bed and all connecting hoses.