Learn More
Biotang Inc Single channel adjustable pipettes 20-200 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 3-year Manufacturer Warranty
Catalog No. 507514928 Shop All BIOTANG Inc Products
Encompass_Preferred
Single channel adjustable pipettes 20-200 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 1-year Manufacturer Warranty
Catalog No. 50-751-4928 Supplier Biotang Inc Supplier No. 770070
Single channel adjustable pipettes 20-200 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 1-year Manufacturer Warranty