Learn More
Biotang Inc Single channel fixed volume pipettes 10 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 3-year Manufacturer Warranty
Catalog No. 507514932 Shop All BIOTANG Inc Products
Encompass_Preferred
Single channel fixed volume pipettes 10 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 1-year Manufacturer Warranty
Catalog No. 50-751-4932 Supplier Biotang Inc Supplier No. 772004
Single channel fixed volume pipettes 10 ul. Easy to calibrate and maintain with calibration tool Included Durable tip cone materials provide excellent chemical resistance. 1-year Manufacturer Warranty