Learn More
Cpi International SGL ELEMENT IRON STD 100ML
SDP
Catalog No. NC9859892
Encompass
Single-Element Standards 1000microgram/mL, Element: Iron, Source: Fe metal, Purity: 99.999%, Matrix: 2% HNO3
Catalog No. NC9859892 Supplier Cpi International Supplier No. S44001000261
Add to Cart
Add to Cart
Single-Element Standards 1000microgram/mL, Element: Iron, Source: Fe metal, Purity: 99.999%, Matrix: 2% HNO3