Learn More
MSC J.T. Eaton Rodenticide Bait Block, Bromethalin, 4 Lb
Catalog No. 502866139
Encompass
J.T. Eaton rodenticide bait blocks are a restricted-use product designed for all-weather application, effectively targeting mice and rats. Ready to use and containing Bromethalin, these solid bait blocks are supplied in a 4 Lb pail.
  • Effectively targets mice and rats
  • Supplied in a pail
  • Contains Bromethalin as active ingredient
  • Solid form, green color, mild odor
  • Insoluble in water
Catalog No. 50-286-6139 Supplier MSC Supplier No. 75041772
J.T. Eaton rodenticide bait blocks are a restricted-use product designed for all-weather application, effectively targeting mice and rats. Ready to use and containing Bromethalin, these solid bait blocks are supplied in a 4 Lb pail.
  • Effectively targets mice and rats
  • Supplied in a pail
  • Contains Bromethalin as active ingredient
  • Solid form, green color, mild odor
  • Insoluble in water