Learn More
Cobex Recorders Inc SGL PEN RECORDER W/INK CHART
SDP
Catalog No. NC2882355
Encompass_Preferred
Customizable 4" single pen recorder with ink chart  Recorder type B Skeleton with doorchart size 4= 4" diameterChart number 4" chart #350Measurement of Pen #1 C= CelsiusInput of pen #1 G= RTD (6"x1/4" stainless steel probe)Measurement of pen #2: 0=No 2nd penMarking system 1= Fiber tip Pen (ink)Voltage:1 = 110 VAC. 50/60 Hz
Catalog No. NC2882355 Supplier Cobex Recorders Inc Supplier No. B4350CF0011Y0
Customizable 4" single pen recorder with ink chart  Recorder type B Skeleton with doorchart size 4= 4" diameterChart number 4" chart #350Measurement of Pen #1 C= CelsiusInput of pen #1 G= RTD (6"x1/4" stainless steel probe)Measurement of pen #2: 0=No 2nd penMarking system 1= Fiber tip Pen (ink)Voltage:1 = 110 VAC. 50/60 Hz