Learn More
J Kem Scientific Inc SGLE ELEM SENSOR TEF 1/4"X24"
SDP
Catalog No. NC2112331
Encompass
T-1/4-T-24 Single-Element Thermocouple TempTemperature sensor - Tdiameter - 1/4 inchCoating - TeflonLength - 24 in
Catalog No. NC2112331 Supplier J Kem Scientific Inc Supplier No. T1/4T24
T-1/4-T-24 Single-Element Thermocouple TempTemperature sensor - Tdiameter - 1/4 inchCoating - TeflonLength - 24 in