Learn More
Aviva Systems Biology Corp SGMS1 Antibody - middle region (ARP55475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SGMS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SGMS1. Alternate names: MGC17342, MOB, MOB1, SMS1, TMEM23, hmob33ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53756