Learn More
Aviva Systems Biology Corp SGMS2 Antibody - N-terminal region (ARP53078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SGMS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SGMS2. Alternate names: MGC26963, SMS2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60914