Learn More
Aviva Systems Biology Corp SGPP1 Antibody - middle region (ARP49967_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49967_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SGPP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SGPP1. Alternate names: SPPase1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-473