Learn More
New England Biolabs, Inc. SgrAI – 1000 units
SDP
Catalog No. 50812328
Encompass
A restriction endonuclease that recognizes the sequence CR^CCGG_YG.
Catalog No. 50-812-328 Supplier New England Biolabs, Inc. Supplier No. R0603S
A restriction endonuclease that recognizes the sequence CR^CCGG_YG.