Learn More
New England Biolabs, Inc. SgrAI – 5000 units
SDP
Catalog No. 50812327
Encompass
A restriction endonuclease that recognizes the sequence CR^CCGG_YG.
Catalog No. 50-812-327 Supplier New England Biolabs, Inc. Supplier No. R0603L
A restriction endonuclease that recognizes the sequence CR^CCGG_YG.